Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-3-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193858
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 843663-18-3
Molecular Formula C7H5BFNO2
Molecular Weight (g/mol) 164.93
MDL Number MFCD03411549
InChI Key DECWLXUOZUMPBF-UHFFFAOYSA-N
PubChem CID 2783130
IUPAC Name (4-cyano-3-fluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=C(C=C1)C#N

Specifications

Color White-Yellow
Quantity 1 g
Synonym 4-cyano-3-fluorobenzeneboronic acid, 4-cyano-3-fluorophenyl boronic acid, 4-borono-2-fluorobenzonitrile, 4-cyano-3-fluoro-phenyl boronic acid, 4-cyano-3-fluoro phenyl boronic acid, 3-fluoro-4-cyanophenylboronic acid, boronic acid, 4-cyano-3-fluorophenyl, 4-cyano-3-fluorophenylboronicacid, pubchem1801, acmc-209pug
Formula Weight 164.93
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-3-fluorophenylboronic Acid (contains varying amounts of Anhydride)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.