Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-4'-methylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. c17415g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 50670-50-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD03703444
InChI Key QIBWMVSMTSYUSK-UHFFFAOYSA-N
PubChem CID 170889
IUPAC Name 4-(4-methylphenyl)benzonitrile
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 110°C
Color White
Quantity 5 g
Synonym 4′C-Methyl-4-biphenylcarbonitrile
Formula Weight 193.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-methylbiphenyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.