Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-4-phenylcyclohexanone 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Plastic bottle
25115-74-6
C13H13NO
199.25
MFCD00044816
GKXOABVSZWCJJK-UHFFFAOYSA-N
4-cyano-4-phenylcyclohexanone,4-oxo-1-phenylcyclohexanecarbonitrile,4-oxo-1-phenylcyclohexanenitrile,cyclohexanecarbonitrile, 4-oxo-1-phenyl,4-oxo-1-phenyl-cyclohexanecarbonitrile,4-oxo-1-phenyl-1-cyclohexanecarbonitrile,acmc-1ciss,4-cyano-4phenylcyclohexanone,4-phenyl 4-cyanocylohexanone,4-phenyl 4-cyanocyclohexanone
91282
4-oxo-1-phenylcyclohexane-1-carbonitrile
C1CC(CCC1=O)(C#N)C2=CC=CC=C2
25115-74-6
C13H13NO
199.25
MFCD00044816
GKXOABVSZWCJJK-UHFFFAOYSA-N
4-cyano-4-phenylcyclohexanone,4-oxo-1-phenylcyclohexanecarbonitrile,4-oxo-1-phenylcyclohexanenitrile,cyclohexanecarbonitrile, 4-oxo-1-phenyl,4-oxo-1-phenyl-cyclohexanecarbonitrile,4-oxo-1-phenyl-1-cyclohexanecarbonitrile,acmc-1ciss,4-cyano-4phenylcyclohexanone,4-phenyl 4-cyanocylohexanone,4-phenyl 4-cyanocyclohexanone
91282
4-oxo-1-phenylcyclohexane-1-carbonitrile
C1CC(CCC1=O)(C#N)C2=CC=CC=C2
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Beige
Quantity 5g
Melting Point 114°C to 119°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5(CN)C6H8(=O)
Solubility Information Solubility in water: .. Other solubilities: soluble in benzene
Formula Weight 199.25
Physical Form Crystalline Solid
Percent Purity ≥96.0%
Chemical Name or Material 4-Cyano-4-phenylcyclohexanone, 97%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 246-631-7