Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanoindole, 99%

Catalog No. p-7054940
Click to view available options
Quantity:
1 g
5 g

CAS: 16136-52-0 | C9H6N2 | 142.161 g/mol

4-Cyanoindole is used as a synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanoindole is used as a synthetic intermediate.

Solubility
Soluble in water.

Notes
Heat, light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16136-52-0
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.161
MDL Number MFCD00152045
InChI Key CEUFGDDOMXCXFW-UHFFFAOYSA-N
Synonym 4-cyanoindole, 4-indolecarbonitrile, indole-4-carbonitrile, indole-4-cyano, 4-cyano-1h-indole, 4-cyano indole, pubchem7327, acmc-209dmi, 1h-indol-4-yl cyanide
PubChem CID 3817602
IUPAC Name 1H-indole-4-carbonitrile
SMILES C1=CC(=C2C=CNC2=C1)C#N

Specifications

Melting Point 120°C to 125°C
Color Gray
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 142.16
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 4-Cyanoindole
Hazard Category H302-H315-H317-H318-H335
Hazard Statement GHS H Statement
H318-H302-H317-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.