Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanophenol, 99%

Catalog No. p-3778009
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 767-00-0 | C7H5NO | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 767-00-0
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.12
MDL Number MFCD00002312
InChI Key CVNOWLNNPYYEOH-UHFFFAOYSA-N
Synonym 4-cyanophenol, p-hydroxybenzonitrile, p-cyanophenol, benzonitrile, 4-hydroxy, benzonitrile, p-hydroxy, 4-cyano phenol, 4-hydroxybenzoic acid nitrile, 4-hydroxy-benzonitrile, 4-hydroxy benzonitrile, 4-hydroxybenzenecarbonitrile
PubChem CID 13019
ChEBI CHEBI:38622
IUPAC Name 4-hydroxybenzonitrile
SMILES C1=CC(=CC=C1C#N)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 110°C to 113°C
Color Beige-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NCC6H4OH
Quantity 25 g
Beilstein 10, 167
Solubility Information Solubility in water: slightly soluble
Formula Weight 119.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanophenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 212-175-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.