Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanophenylhydrazine hydrochloride, 97+%

Catalog No. p-3593329
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 2863-98-1 | C7H7N3 | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2863-98-1
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00673994
InChI Key DZUUSHCOMPROCJ-UHFFFAOYSA-N
Synonym 4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j
PubChem CID 16212962
SMILES NNC1=CC=C(C=C1)C#N

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 249.0°C
Color Brown to Orange
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Linear Formula NCC6H4NHNH2·HCl
Quantity 1 g
Solubility Information Solubility in water: soluble
IUPAC Name 4-hydrazinylbenzonitrile;hydrochloride
Formula Weight 169.61
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 4-Cyanophenylhydrazine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fre

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.