Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanopyridine, 98%

Catalog No. AAA1028636
Click to view available options
Quantity:
10 g
100 g
500 g
2500 g

CAS: 100-48-1 | C6H4N2 | 104.112 g/mol

4-Cyanopyridine is used as an intermediate in organic synthesis and pharmaceutical substances like isonicotinylhydrazide, which is used in the treatment of tuberculosis. It is used as a precursor for the preparation of isonicotinic acid and 4-diemthylaminopyridine (DMAP). It is involved in the synthesis of 6-methyl-2-pyridin-4-yl-pyrimidin-4-ylamine by reacting with acetonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanopyridine is used as an intermediate in organic synthesis and pharmaceutical substances like isonicotinylhydrazide, which is used in the treatment of tuberculosis. It is used as a precursor for the preparation of isonicotinic acid and 4-diemthylaminopyridine (DMAP). It is involved in the synthesis of 6-methyl-2-pyridin-4-yl-pyrimidin-4-ylamine by reacting with acetonitrile.

Solubility
Soluble in water.

Notes
Incompatible with strong acids, strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-48-1
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.112
MDL Number MFCD00006417
InChI Key GPHQHTOMRSGBNZ-UHFFFAOYSA-N
Synonym 4-cyanopyridine, isonicotinonitrile, 4-pyridinecarbonitrile, isonicotinic acid nitrile, 4-pyridinenitrile, 4-cyano pyridine, .gamma.-cyanopyridine, gamma-cyanopyridine, p-cyanopyridine, pubchem11155
PubChem CID 7506
ChEBI CHEBI:28020
IUPAC Name pyridine-4-carbonitrile
SMILES C1=CN=CC=C1C#N

Specifications

Melting Point 76°C to 80°C
pH 6
Boiling Point 194°C to 196°C
Flash Point 88°C (190°F)
Odor Pungent
Quantity 500 g
Beilstein 107712
Solubility Information Soluble in water.
Formula Weight 104.11
Percent Purity 98%
Chemical Name or Material 4-Cyanopyridine
Show More Show Less
Hazard Category H227-H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 202-856-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.