Learn More
4-Cyanostyrene, 97%, stab. with 0.05% 4-tert-butyl catechol
Description
4-Cyanostyrene is used as an intermediate in organic synthesis. It is used in the preparation of poly(4-cyanostyrene) polymer , which is a macromolecular device having nanoscale order and finds a wide range of potential applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Cyanostyrene is used as an intermediate in organic synthesis. It is used in the preparation of poly(4-cyanostyrene) polymer , which is a macromolecular device having nanoscale order and finds a wide range of potential applications.
Solubility
Immiscible with water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with polymerizing initiators, polymerization catalysts and strong oxidizing agents.
Specifications
Specifications
| Boiling Point | 92°C to 93°C (3 mmHg) |
| Refractive Index | 1.5775 |
| Quantity | 5 g |
| UN Number | UN3276 |
| Synonym | 4-cyanostyrene, 4-vinylbenzonitrile, benzonitrile, 4-ethenyl, p-cyanostyrene, 4-vinyl-benzonitrile, benzonitrile, p-vinyl, p-vinylbenzonitrile, acmc-20amk5, benzonitrile,4-ethenyl, 4-vinylbenzenecarbonitrile |
| Solubility Information | Immiscible with water. |
| Formula Weight | 129.16 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Cyanostyrene, stabilized with 0.05% 4-tert-butyl catechol |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H331-H302-H312-H315-H319-H335 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.