Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyclohexylbenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. C152325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C152325G 25 g
C15235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C152325G Supplier TCI America Supplier No. C152325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20029-52-1
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00039459
InChI Key QCIWHVKGVVQHIY-UHFFFAOYSA-N
Synonym 4-cyclohexyl-benzoic acid, benzoic acid, 4-cyclohexyl, 4cyclohexylbenzoic acid, 4-cyclohexylbenzoicacid, p-cyclohexylbenzoic acid, acmc-1cmbu, cyclohexybenzoicacid, 4-carboxyphenyl cyclohexane
PubChem CID 88337
IUPAC Name 4-cyclohexylbenzoic acid
SMILES C1CCC(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 200°C
Color White
Quantity 25 g
Formula Weight 204.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyclohexylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.