Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Cyclohexylmethylsulfamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog No. p-7036229
Click to view available options
Quantity:
250 mg
1 g

Catalog No. AAH54490MD

Supplier: Thermo Scientific Chemicals H54490MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1221823-27-3
Molecular Formula C13H20BNO4S
Molecular Weight (g/mol) 297.176
MDL Number MFCD20265279
InChI Key JDFPAMLPRYKLRV-UHFFFAOYSA-N
Synonym 4-cyclohexylmethylsulfamoyl benzeneboronic acid, 4-cyclohexylmethylsulfamoyl phenylboronic acid
PubChem CID 57655465
IUPAC Name [4-(cyclohexylmethylsulfamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)S(=O)(=O)NCC2CCCCC2)(O)O

Specifications

Quantity 250 mg
Formula Weight 297.18
Percent Purity 97%
Chemical Name or Material 4-(Cyclohexylmethylsulfamoyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.