Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 4-DAMP
GREENER_CHOICESDP

Catalog No. 048250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-825-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 04-825-0 Supplier Tocris Bioscience™ Supplier No. 0482/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Muscarinic M3 antagonist

Antagonist at the M3 cholinergic receptor. [3H]-4-DAMP selectively labels M1 and M3 receptors.

Chemical Identifiers

CAS 1952-15-4
Molecular Formula C21H26INO2

Specifications

Quantity 50 mg
Formula Weight 451.33
Purity >99%
Molecular Weight (g/mol) 451.348
Chemical Name or Material 4-DAMP
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.