Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Diethylamino-2-butyn-1-ol 95.0+%, TCI America™
SDP

Catalog No. D280525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D280525ML 25 mL
D28055ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D280525ML Supplier TCI America Supplier No. D280525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10575-25-4
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.21
MDL Number MFCD00671356
InChI Key ACGZBRWTWOZSFU-UHFFFAOYSA-N
Synonym 4-diethylamino but-2-yn-1-ol, 4-diethylamino-2-butyn-1-ol, unii-ma2de62u5w, ma2de62u5w, 2-butyn-1-ol,4-diethylamino, 2-butyn-1-ol, 4-diethylamino, acmc-2098hk, 4-diethylamino-2-butyne-ol, 1-diethylaminobut-2-yn-4-ol
PubChem CID 82735
IUPAC Name 4-(diethylamino)but-2-yn-1-ol
SMILES CCN(CC)CC#CCO

Specifications

Quantity 25 mL
Formula Weight 141.21
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Diethylamino-2-butyn-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.