Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Diethylamino)salicylaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
50g
Packaging:
Glass bottle
17754-90-4
C11H15NO2
193.24
XFVZSRRZZNLWBW-UHFFFAOYSA-N
4-diethylamino salicylaldehyde,4-diethylamino-2-hydroxybenzaldehyde,4-diethylaminosalicylaldehyde,4-n,n-diethylamino salicylaldehyde,benzaldehyde, 4-diethylamino-2-hydroxy,p-diethylamino salicylaldehyde,unii-ou5nfu4681,2-hydroxy-4-diethylaminobenzaldehyde,4-n,n-diethylaminosalicylic aldehyde,4-diethylamino-2-hydroxy-benzaldehyde
87293
4-(diethylamino)-2-hydroxybenzaldehyde
CCN(CC)C1=CC(=C(C=C1)C=O)O
17754-90-4
C11H15NO2
193.24
XFVZSRRZZNLWBW-UHFFFAOYSA-N
4-diethylamino salicylaldehyde,4-diethylamino-2-hydroxybenzaldehyde,4-diethylaminosalicylaldehyde,4-n,n-diethylamino salicylaldehyde,benzaldehyde, 4-diethylamino-2-hydroxy,p-diethylamino salicylaldehyde,unii-ou5nfu4681,2-hydroxy-4-diethylaminobenzaldehyde,4-n,n-diethylaminosalicylic aldehyde,4-diethylamino-2-hydroxy-benzaldehyde
87293
4-(diethylamino)-2-hydroxybenzaldehyde
CCN(CC)C1=CC(=C(C=C1)C=O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 60.0°C to 64.0°C
Color Brown to Purple
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 14, 234
Formula Weight 193.24
Physical Form Powder or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-(Diethylamino)salicylaldehyde, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 241-745-3

RTECSNumber : CU5615150

TSCA : TSCA