Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Diethylamino)salicylaldehyde, 98%

Catalog No. p-108063
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 17754-90-4 | C11H15NO2 | 193.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17754-90-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.24
InChI Key XFVZSRRZZNLWBW-UHFFFAOYSA-N
Synonym 4-diethylamino salicylaldehyde, 4-diethylamino-2-hydroxybenzaldehyde, 4-diethylaminosalicylaldehyde, 4-n,n-diethylamino salicylaldehyde, benzaldehyde, 4-diethylamino-2-hydroxy, p-diethylamino salicylaldehyde, unii-ou5nfu4681, 2-hydroxy-4-diethylaminobenzaldehyde, 4-n,n-diethylaminosalicylic aldehyde, 4-diethylamino-2-hydroxy-benzaldehyde
PubChem CID 87293
IUPAC Name 4-(diethylamino)-2-hydroxybenzaldehyde
SMILES CCN(CC)C1=CC(=C(C=C1)C=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 60.0°C to 64.0°C
Color Brown to Purple
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 14, 234
Formula Weight 193.24
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-(Diethylamino)salicylaldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-745-3
RTECSNumber CU5615150
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.