Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester, 96%

Catalog No. AAH5988003
Click to view available options
Quantity:
250 mg
1 g

CAS: 1628442-55-6 | C13H16BF3O3 | 288.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1628442-55-6
Molecular Formula C13H16BF3O3
Molecular Weight (g/mol) 288.073
MDL Number MFCD22419269
InChI Key LLJRSXMGPKHUQX-UHFFFAOYSA-N
Synonym 2-4-difluoromethoxy-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester
PubChem CID 79661309
IUPAC Name 2-[4-(difluoromethoxy)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)OC(F)F)F

Specifications

Quantity 1 g
Formula Weight 288.07
Percent Purity 96%
Chemical Name or Material 4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.