Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-(Difluoromethoxy)acetophenone 98.0+%, TCI America™
SDP

Catalog No. D47601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47601G 1 g
D47605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47601G Supplier TCI America Supplier No. D47601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 83882-67-1
Molecular Formula C9H8F2O2
Molecular Weight (g/mol) 186.158
MDL Number MFCD00042250
InChI Key GIGWRVLNOYPOIT-UHFFFAOYSA-N
PubChem CID 737356
IUPAC Name 1-[4-(difluoromethoxy)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)OC(F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 186.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-(Difluoromethoxy)acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.