Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)benzoyl chloride, 95%, Thermo Scientific™

Catalog No. AC399690010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4755-50-4
InChI Key UGJDXRVQCYBXAJ-UHFFFAOYSA-N
Synonym 4-dimethylamino benzoyl chloride, benzoyl chloride, 4-dimethylamino, 4-dimethylamino benzoylchloride, 4-dimethylaminobenzoylchloride, acmc-20amjv, p-dimethylaminobenzoylchlorid, 4-dimethyaminobenzoyl chloride, p-dimethylaminobenzoyl chloride, 4-dimethlamino benzoyl chloride
PubChem CID 4400782
IUPAC Name 4-(dimethylamino)benzoyl chloride
SMILES CN(C)C1=CC=C(C=C1)C(=O)Cl

Specifications

Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C9H10ClNO
Linear Formula (CH3)2NC6H4COCl
Quantity 1 g
Molecular Weight (g/mol) 183.635
Formula Weight 183.64
Chemical Name or Material 4-(Dimethylamino)benzoyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.