Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)benzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. D348225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D348225G 25 g
D34825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D348225G Supplier TCI America Supplier No. D348225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1703-46-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD03840449
InChI Key WQBCAASPALGAKX-UHFFFAOYSA-N
Synonym 4-Hydroxymethyl-N,N-dimethylaniline
PubChem CID 137166
IUPAC Name [4-(dimethylamino)phenyl]methanol
SMILES CN(C)C1=CC=C(CO)C=C1

Specifications

Color Yellow
Boiling Point 123°C
Quantity 25 g
Formula Weight 151.21
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 4-(Dimethylamino)benzyl Alcohol
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.