Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)cinnamic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1552-96-1
C11H13NO2
191.23
MFCD00004397
CQNPVMCASGWEHM-VMPITWQZSA-N
4-dimethylamino cinnamic acid,4-dimethylaminocinnamic acid,p-dimethylamino cinnamic acid,3-4-dimethylamino phenyl acrylic acid,4-n,n-dimethylamino cinnamic acid,2e-3-4-dimethylamino phenyl prop-2-enoic acid,e-3-4-dimethylaminophenyl prop-2-enoic acid,e-3-4-dimethylamino phenyl acrylic acid
1540638
(E)-3-[4-(dimethylamino)phenyl]prop-2-enoic acid
CN(C)C1=CC=C(C=C1)C=CC(=O)O
1552-96-1
C11H13NO2
191.23
MFCD00004397
CQNPVMCASGWEHM-VMPITWQZSA-N
4-dimethylamino cinnamic acid,4-dimethylaminocinnamic acid,p-dimethylamino cinnamic acid,3-4-dimethylamino phenyl acrylic acid,4-n,n-dimethylamino cinnamic acid,2e-3-4-dimethylamino phenyl prop-2-enoic acid,e-3-4-dimethylaminophenyl prop-2-enoic acid,e-3-4-dimethylamino phenyl acrylic acid
1540638
(E)-3-[4-(dimethylamino)phenyl]prop-2-enoic acid
CN(C)C1=CC=C(C=C1)C=CC(=O)O
Packaging Glass bottle
Melting Point 225°C to 228°C
Color Yellow to Beige
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2NC6H4CH=CHCO2H
Beilstein 14, 522
Formula Weight 191.23
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-(Dimethylamino)cinnamic acid, 99%
Show More Show Less

EINECSNumber : 216-299-8