Learn More
4-(Dimethylamino)pyridine, 99%
Description
4-(Dimethylamino)pyridine is a useful nucleophilic catalyst for various reactions viz. esterifications with anhydrides, the Baylis-Hillman reaction, hydrosilylations, tritylation and Steglich rearrangement. It is also an effective catalyst for transesterification of beta-keto esters and silylation of alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-(Dimethylamino)pyridine is a useful nucleophilic catalyst for various reactions viz. esterifications with anhydrides, the Baylis-Hillman reaction, hydrosilylations, tritylation and Steglich rearrangement. It is also an effective catalyst for transesterification of beta-keto esters and silylation of alcohols.
Solubility
Soluble in methanol, benzene, ethyl acetate, chloroform, methylene chloride, acetone, and acetic acid. Slightly soluble in ether, diisopropyl ether, cyclohexane, hexane, and water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
Specifications
| Melting Point | 110°C to 113°C |
| Boiling Point | 162°C (50 mmHg) |
| Flash Point | 124°C (255°F) |
| Assay Percent Range | 99% |
| Quantity | 25 g |
| UN Number | UN2928 |
| Beilstein | 110354 |
| Solubility Information | Soluble in methanol,benzene,ethyl acetate,chloroform,methylene chloride,acetone,and acetic acid. Slightly soluble in ether,diisopropyl ether,cyclohexane,hexane,and water. |
| Formula Weight | 122.17 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H301+H331-H310-H315-H318-H370 |
| Hazard Statement | GHS H Statement H301-H310-H314-H318 |
| Precautionary Statement | P260-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P307+P311-P310-P311-P330-P332+P313-P361-P363-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: II |
| EINECSNumber | 214-353-5 |
| RTECSNumber | US9230000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.