Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminobenzaldehyde, 98%

Catalog No. p-7081655
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 100-10-7 | C9H11NO | 149.19 g/mol

4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

Solubility
Soluble in alcohol, ether, chloroform, acetic acid. Insoluble in water.

Notes
Light sensitive. Stable at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Melting Point 73°C to 75°C
Boiling Point 176°C to 177°C (17 mmHg)
Flash Point 164°C (327°F)
Odor Strong
Quantity 100 g
Beilstein 606802
Sensitivity Air and light sensitive
Merck Index 14,3230
Solubility Information Soluble in alcohol,ether,chloroform,acetic acid. Insoluble in water.
Formula Weight 149.19
Percent Purity 98%
Chemical Name or Material 4-Dimethylaminobenzaldehyde
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 202-819-0
RTECSNumber CU5775000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.