Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Dimethylaminobenzaldehyde, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 70°C to 75°C |
| Color | White to Off-white |
| Density | 5.15 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Linear Formula | (CH3)2NC6H4CHO |
| Quantity | 500 g |
| Beilstein | 14, 31 |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: WARNING! |
| Hazard Statement | GHS H Statement: Harmful to aquatic life with long lasting effects. Harmful if swallowed. |
| Precautionary Statement | Emergency Overview Harmful if swallowed. May cause eye, skin, and respiratory tract irritation . Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. |
| EINECSNumber | 202-819-0 |
| RTECSNumber | CU5775000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction