Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Dimethylaminobutyraldehyde diethyl acetal, 97%

Click to view available options
Quantity:
25g
100g
500g
1116-77-4
C10H23NO2
189.299
MFCD00671479
QKXMWBLNSPNBEY-UHFFFAOYSA-N
4,4-diethoxy-n,n-dimethyl-1-butanamine,4,4-diethoxy-butyl-dimethyl-amine,4,4-diethoxy-n,n-dimethylbutylamine,4,4-diethoxy-n,n-dimethylbutanamine,4,4-diethoxy-n,n-dimethyl butylamine,4-n,n-dimethylamino butanal diethyl acetal,4,4-diethoxy-n, n-dimethyl-butylamine,4,4-diethoxy-n,n-dimethyl-butan-1-amine,4-dimethylaminobutyraldehyde diethyl acetal,n,n-dimethyl-4-aminobutanal diethyl acetal
547078
4,4-diethoxy-N,N-dimethylbutan-1-amine
CCOC(CCCN(C)C)OCC
1116-77-4
C10H23NO2
189.299
MFCD00671479
QKXMWBLNSPNBEY-UHFFFAOYSA-N
4,4-diethoxy-n,n-dimethyl-1-butanamine,4,4-diethoxy-butyl-dimethyl-amine,4,4-diethoxy-n,n-dimethylbutylamine,4,4-diethoxy-n,n-dimethylbutanamine,4,4-diethoxy-n,n-dimethyl butylamine,4-n,n-dimethylamino butanal diethyl acetal,4,4-diethoxy-n, n-dimethyl-butylamine,4,4-diethoxy-n,n-dimethyl-butan-1-amine,4-dimethylaminobutyraldehyde diethyl acetal,n,n-dimethyl-4-aminobutanal diethyl acetal
547078
4,4-diethoxy-N,N-dimethylbutan-1-amine
CCOC(CCCN(C)C)OCC
Quantity 500g
Flash Point 70°C (158°F)
Formula Weight 189.3
Percent Purity 97%
Chemical Name or Material 4-Dimethylaminobutyraldehyde diethyl acetal

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning