Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminocinnamaldehyde 98.0+%, TCI America™
SDP

Catalog No. D064825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D064825G 25 g
D06485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D064825G Supplier TCI America Supplier No. D064825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6203-18-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00007002
InChI Key RUKJCCIJLIMGEP-ONEGZZNKSA-N
Synonym 4-dimethylamino cinnamaldehyde, 4-dimethylaminocinnamaldehyde, p-dimethylaminocinnamaldehyde, 3-4-dimethylamino phenyl acrylaldehyde, 4-dimethylcinnamaldehyde, p-dimethylaminocinnamic aldehyde, p-dimethylamino cinnamaldehyde, cinnamaldehyde, p-dimethylamino, unii-9rsi7wz9f0, dmaca reagent
PubChem CID 5284506
IUPAC Name (E)-3-[4-(dimethylamino)phenyl]prop-2-enal
SMILES CN(C)C1=CC=C(C=C1)C=CC=O

Specifications

Melting Point 140°C
Quantity 25 g
Formula Weight 175.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminocinnamaldehyde
EINECSNumber (3)-1187
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.