Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminopyridinium Bromide Perbromide 97.0+%, TCI America™
SDP

Catalog No. D178725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D178725G 25 g
D17875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D178725G Supplier TCI America Supplier No. D178725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92976-81-3
Molecular Formula C7H10Br3N2-
Molecular Weight (g/mol) 361.883
MDL Number MFCD00043439
InChI Key TYDZHQMKSLAFKM-UHFFFAOYSA-N
Synonym 4-dimethylamino pyridine hydrobromide perbromide
PubChem CID 131863682
SMILES CN(C)C1=CC=NC=C1.Br[Br-]Br

Specifications

Melting Point 131°C
Quantity 25 g
UN Number 3261
Formula Weight 362.89
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminopyridinium Bromide Perbromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.