Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Diphenylaminobenzaldehyde, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4181-05-9
C19H15NO
273.33
MFCD00145131
UESSERYYFWCTBU-UHFFFAOYSA-N
4-diphenylamino benzaldehyde,4-n,n-diphenylamino benzaldehyde,4-diphenylaminobenzaldehyde,4-diphenylamino-benzaldehyde,benzaldehyde, 4-diphenylamino,4-formyltriphenylamine,p-formyltriphenylamine,4-n-phenylanilino benzaldehyde,p-diphenylaminobenzaldehyde,4-n,n-diphenylamino-benzaldehyde
77846
4-(N-phenylanilino)benzaldehyde
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O
4181-05-9
C19H15NO
273.33
MFCD00145131
UESSERYYFWCTBU-UHFFFAOYSA-N
4-diphenylamino benzaldehyde,4-n,n-diphenylamino benzaldehyde,4-diphenylaminobenzaldehyde,4-diphenylamino-benzaldehyde,benzaldehyde, 4-diphenylamino,4-formyltriphenylamine,p-formyltriphenylamine,4-n-phenylanilino benzaldehyde,p-diphenylaminobenzaldehyde,4-n,n-diphenylamino-benzaldehyde
77846
4-(N-phenylanilino)benzaldehyde
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O
Quantity 25g
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 129°C to 133°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula (C6H5)2NC6H4C(=O)H
Formula Weight 273.33
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4-Diphenylaminobenzaldehyde
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning