Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dodecyldiethylenetriamine (Tech.), 90%, Thermo Scientific™

Catalog No. AC338050250
Click to view available options
Quantity:
25 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4182-44-9
Molecular Formula C16H37N3
Molecular Weight (g/mol) 271.493
InChI Key BRNPMBFYMPUNSK-UHFFFAOYSA-N
Synonym 4-dodecyldiethylenetriamine, unii-vg94uin7ys, 1,2-ethanediamine, n-2-aminoethyl-n-dodecyl, n-2-aminoethyl-n-dodecyl-1,2-ethanediamine, vg94uin7ys, n1-2-aminoethyl-n1-dodecyl-1,2-ethanediamine, 1,2-ethanediamine, n1-2-aminoethyl-n1-dodecyl, n-2-aminoethyl-n-dodecylethylenediamine, bis 2-aminoethyl dodecyl amine, n'-2-aminoethyl-n'-dodecylethane-1,2-diamine
PubChem CID 77847
IUPAC Name N'-(2-aminoethyl)-N'-dodecylethane-1,2-diamine
SMILES CCCCCCCCCCCCN(CCN)CCN

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Refractive Index 1.4640 to 1.4680 (20°C, 589nm)
Linear Formula CH3(CH2)11N(CH2CH2NH2)2
Quantity 25 g
Formula Weight 271.48
Percent Purity ≥88%
Grade Technical
Physical Form Liquid
Chemical Name or Material 4-Dodecyldiethylenetriamine, tech., 90%
Show More Show Less
Hazard Category Danger
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.