Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Epianhydrochlortetracycline hydrochloride, can be used as secondary standard, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 222.0°C |
| Color | Orange |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 80% min. (as is,calculated) |
| Packaging | Glass bottle |
| Quantity | 100 mg |
| Solubility Information | Solubility in water: 10g/L. Other solubilities: 29g/L methanol |
| Formula Weight | 497.33 |
| Percent Purity | ≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC) |
| Physical Form | Powder |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: Harmful if swallowed. Causes serious eye irritation. Suspected of damaging the unborn child. |
| Precautionary Statement | GHS P Statement: Use personal protective equipment as required. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction