Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Ethoxyacetophenone, 98%

Catalog No. AAA1103114
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1676-63-7 | C10H12O2 | 164.20 g/mol

4'-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.

Solubility
Soluble in alcohol, water(791.1 mg/L).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1676-63-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00009095
InChI Key YJFNFQHMQJCPRG-UHFFFAOYSA-N
Synonym 4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone
PubChem CID 72872
IUPAC Name 1-(4-ethoxyphenyl)ethanone
SMILES CCOC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 35°C to 38°C
Boiling Point 145°C to 146°C (13 mmHg)
Flash Point 134°C (273°F)
Quantity 25 g
Beilstein 636783
Solubility Information Soluble in alcohol,water(791.1mg/L).
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 4'-Ethoxyacetophenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-825-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.