Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog No. AAH5508703
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 851334-92-4
Molecular Formula C15H20BFO4
Molecular Weight (g/mol) 294.129
MDL Number MFCD18383988
InChI Key ZXSGWPHDXQZIAF-UHFFFAOYSA-N
Synonym ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-ethoxycarbonyl-2-fluorophenylboronic acid, pinacol ester, 4-ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzoate, ethyl 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
PubChem CID 23134230
IUPAC Name ethyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OCC)F

Specifications

Quantity 1 g
Beilstein 14562502
Formula Weight 294.13
Percent Purity 97%
Chemical Name or Material 4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.