Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-2-isocyanatobenzoate, 97%

Catalog No. AC440150250
Click to view available options
Quantity:
5 g
25 g

Catalog No. AC440150250

Supplier: Thermo Scientific Chemicals 440150250

CAS: 1793-07-3 | C9H7NO3 | 177.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1793-07-3
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
InChI Key ATFKBWNEVZUSKC-UHFFFAOYSA-N
Synonym 2-carbomethoxyphenyl isocyanate, 2-methoxycarbonylphenyl isocyanate, 2-methoxycarbonyl phenyl isocyanate, 2-carbomethoxyphenylisocyanate, 2-methoxycarbonyl phenylisocyanate, benzoic acid, 2-isocyanato-, methyl ester, acmc-1bqmd, methyl 2 isocyanatobenzoate, methyl-2-isocyanatobenzoate, #
PubChem CID 519494
IUPAC Name methyl 2-isocyanatobenzoate
SMILES COC(=O)C1=CC=CC=C1N=C=O

Specifications

Melting Point 47.0°C to 50.0°C
Color White
Boiling Point 99.0°C (0.5 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 g
Formula Weight 177.16
Percent Purity 97%
Physical Form Crystalline Solid or Solid
Chemical Name or Material Methyl-2-isocyanatobenzoate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic if inhaled.
May cause allergy or asthma symptoms
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.