Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethynylbenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. E11301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11301G 1 g
E1130200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11301G Supplier TCI America Supplier No. E11301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1788-08-5
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD11040256
InChI Key OQPUCENNUFNCQO-UHFFFAOYSA-N
PubChem CID 44457221
IUPAC Name 4-ethynylbenzenesulfonamide
SMILES C#CC1=CC=C(C=C1)S(=O)(=O)N

Specifications

Melting Point 180°C
Quantity 1 g
Formula Weight 181.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethynylbenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.