Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-methoxyphenylboronic acid, 97%, Thermo Scientific™

Catalog No. AC434080010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 179899-07-1
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
MDL Number MFCD03788423
InChI Key ADJBXDCXYMCCAD-UHFFFAOYSA-N
Synonym 4-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-4-fluorophenylboronic acid, 4-fluoro-2-methoxybenzeneboronic acid, 4-fluoro-2-methoxy-phenyl boronic acid, 4-fluoro-2-methoxyphenyl-boronic acid, 5-fluoroanisole-2-boronic acid, 4-fluoro-2-methoxy phenyl boronic acid, boronic acid, 4-fluoro-2-methoxyphenyl, pubchem19716, acmc-1c2tm
PubChem CID 2782672
IUPAC Name (4-fluoro-2-methoxyphenyl)boronic acid
SMILES COC1=CC(F)=CC=C1B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 133.9°C to 137.9°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 169.95
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-methoxyphenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.