Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-methylbenzaldehyde 95.0+%, TCI America™
SDP

Catalog No. F07931G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 63082-45-1
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.141
MDL Number MFCD02261764
InChI Key ADCFIKGEGWFWEA-UHFFFAOYSA-N
Synonym 4-fluoro-o-tolualdehyde, 4-fluoro-2-methyl benzaldehyde, 2-methyl-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-2-methyl, 4-fluoro-2-methyl-benzaldehyde, pubchem1456, 5-fluoro-2-formyltoluene, acmc-1b7wn, ksc352s3j, 4-fluoro-2-methylbenzaldehyde?
PubChem CID 2783217
IUPAC Name 4-fluoro-2-methylbenzaldehyde
SMILES CC1=C(C=CC(=C1)F)C=O

Specifications

Color Yellow
Boiling Point 55°C
Quantity 1 g
Formula Weight 138.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-methylbenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.