Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-methylbenzonitrile 99.0+%, TCI America™
SDP

Catalog No. F09621G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 147754-12-9
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD03095106
InChI Key BJBXUIUJKPOZLV-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorobenzonitrile, 4-fluoro-2-methyl-benzonitrile, benzonitrile, 4-fluoro-2-methyl, 2-cyano-5-fluorotoluene, 4-fluoro-2-methylbenzenecarbonitrile, pubchem1554, intermediates-zcf02129, methyl 4-fluorobenzonitrile, 4-fuoro-2-methylbenzonitrile, ksc174s6l
PubChem CID 3826055
IUPAC Name 4-fluoro-2-methylbenzonitrile
SMILES CC1=C(C=CC(=C1)F)C#N

Specifications

Melting Point 71°C
Color White
Quantity 1 g
UN Number 3439
Formula Weight 135.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-methylbenzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.