Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-methylbenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. F111625G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 455-84-5
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.583
MDL Number MFCD01631426
InChI Key FMFLKTDRPXYUGP-UHFFFAOYSA-N
Synonym 4-fluoro-3-methylbenzoylchloride, 4-fluoro-m-toluoyl chloride, 4-fluoro-3-methyl-benzoyl chloride, benzoyl chloride, 4-fluoro-3-methyl, acmc-1arwr, 5-chlorocarbonyl-2-fluorotoluene, 4-fluoranyl-3-methyl-benzoyl chloride, 4-fluoro-3-methyl benzoyl chloride, 4-fluoro-3-methyl-benzoic acid chloride, 4-fluoro-3-methylbenzoyl chloride
PubChem CID 2774589
IUPAC Name 4-fluoro-3-methylbenzoyl chloride
SMILES CC1=C(C=CC(=C1)C(=O)Cl)F

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
UN Number 3265
Formula Weight 172.58
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-methylbenzoyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.