Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-nitroanisole, 95%

Catalog No. AAH61554MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61554MD 250 mg
AAH6155403 1 g
AAH6155406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH61554MD Supplier Thermo Scientific Chemicals Supplier No. H61554MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

CAS: 61324-93-4 | C7H6FNO3 | 171.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61324-93-4
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.127
MDL Number MFCD11848442
InChI Key ZRIKJXDEJYMBEJ-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitroanisole, benzene, 1-fluoro-4-methoxy-2-nitro, 3-fluoro-4-nitrobenzene,-1-methoxy, ksc613a5f, 2-fluoro-5-methoxynitrobenzene, 2-fluoro-5-methoxy-nitrobenzene, #
PubChem CID 565653
IUPAC Name 1-fluoro-4-methoxy-2-nitrobenzene
SMILES COC1=CC(=C(C=C1)F)[N+](=O)[O-]

Specifications

Quantity 250 mg
Formula Weight 171.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Fluoro-3-nitroanisole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.