Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-4'-hydroxybenzophenone 98.0+%, TCI America™
SDP

Catalog No. F026910G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 25913-05-7
Molecular Formula C13H9FO2
Molecular Weight (g/mol) 216.211
MDL Number MFCD00011484
InChI Key HLRVUOFDBXRZBI-UHFFFAOYSA-N
PubChem CID 117666
IUPAC Name (4-fluorophenyl)-(4-hydroxyphenyl)methanone
SMILES C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)F)O

Specifications

Melting Point 168°C
Quantity 10 g
Formula Weight 216.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-4′-hydroxybenzophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.