Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Fluoro-7-Nitrobenzofurazan Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $499.50
Specifications
Antigen | 4-Fluoro-7-Nitrobenzofurazan |
---|---|
Concentration | 5.0 mg/mL |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017457
![]() |
Novus Biologicals
NB100622850.1ML |
0.1 mL |
Each for $207.00
|
|
|||||
NB10062285
![]() |
Novus Biologicals
NB10062285 |
0.5 mL |
Each for $499.50
|
|
|||||
Description
4-Fluoro-7-Nitrobenzofurazan Polyclonal specifically detects 4-Fluoro-7-Nitrobenzofurazan in Non-species specific samples. It is validated for ELISA.Specifications
4-Fluoro-7-Nitrobenzofurazan | |
ELISA | |
Unconjugated | |
Rabbit | |
Non-species specific | |
IgG | |
Protein G purified | |
NB100-62285 is specific for 4-fluoro-7-nitrobenzofurazan (NBD), reacting with both free and conjugated forms of NBD. |
5.0 mg/mL | |
Polyclonal | |
Purified | |
RUO | |
NBD-F conjugated to cationised BSA. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title