Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Fluoroacetanilide 98.0+%, TCI America™
SDP

Catalog No. f052125g
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 351-83-7
Molecular Formula C8H8FNO
Molecular Weight (g/mol) 153.156
InChI Key JHEFOJNPLXSWNZ-UHFFFAOYSA-N
Synonym 4-fluoroacetanilide, n-4-fluorophenyl acetamide, 4'-fluoroacetanilide, p-fluoroacetanilide, acetanilide, 4'-fluoro, acetamide, n-4-fluorophenyl, n-p-fluorophenylacetamide, 1-acetamido-4-fluorobenzene, 4-fluorophenyl acetamide, acetamide,n-4-fluorophenyl
PubChem CID 9601
IUPAC Name N-(4-fluorophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)F

Specifications

Melting Point 154°C
Color White
Quantity 25 g
Formula Weight 153.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Fluoroacetanilide
RTECSNumber AE2977000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.