Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoroaniline 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
371-40-4
C6H6FN
111.12
MFCD00007829
KRZCOLNOCZKSDF-UHFFFAOYSA-N
p-fluoroaniline,benzenamine, 4-fluoro,1-amino-4-fluorobenzene,4-fluorobenzenamine,p-fluorophenylamine,4-fluoronaniline,aniline, p-fluoro,aniline, 4-fluoro,4-fluoranilin,para-fluoroaniline
9731
CHEBI:28546
4-fluoroaniline
C1=CC(=CC=C1N)F
371-40-4
C6H6FN
111.12
MFCD00007829
KRZCOLNOCZKSDF-UHFFFAOYSA-N
p-fluoroaniline,benzenamine, 4-fluoro,1-amino-4-fluorobenzene,4-fluorobenzenamine,p-fluorophenylamine,4-fluoronaniline,aniline, p-fluoro,aniline, 4-fluoro,4-fluoranilin,para-fluoroaniline
9731
CHEBI:28546
4-fluoroaniline
C1=CC(=CC=C1N)F
Specific Gravity 1.157
Density 1.1570g/mL
Flash Point 73°C
Melting Point -1.9°C
Color Undesignated
Quantity 25mL
Packaging Glass bottle
Boiling Point 187.0°C
Refractive Index 1.5385 to 1.5405
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula FC6H4NH2
Beilstein 12, 597
Merck Index 15, 4200
Solubility Information Solubility in water: 33g/L (20°C). Other solubilities: soluble in acetone,ethanol,xylene alcohol,,carbon tetrachloride and diethyl ether
Formula Weight 111.12
Physical Form Oily Liquid
Percent Purity 98%
Chemical Name or Material 4-Fluoroaniline, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed. Causes severe skin burns and eye damage. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

EINECSNumber : 206-735-5

RTECSNumber : BY1575000

TSCA : TSCA