Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Fluorotoluene, 99%

Catalog No. AC119665000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 95-52-3 | C7H7F | 110.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-52-3
MDL Number MFCD00000322
InChI Key MMZYCBHLNZVROM-UHFFFAOYSA-N
Synonym 2-fluorotoluene, o-fluorotoluene, benzene, 1-fluoro-2-methyl, toluene, o-fluoro, 1-methyl-2-fluorobenzene, unii-l55xkb3k7h, 1-fluoro-2-methyl-benzene, ortho-fluorotoluene, fluorotoluene, l55xkb3k7h
PubChem CID 7241
IUPAC Name 1-fluoro-2-methylbenzene
SMILES CC1=CC=CC=C1F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -62.0°C
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 113.0°C to 114.0°C
Flash Point 8°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7F
Refractive Index 1.4720 to 1.4750
Linear Formula CH3C6H4F
Quantity 500 g
Beilstein 05, 290
Specific Gravity 1
Merck Index 15, 4212
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Molecular Weight (g/mol) 110.13
Formula Weight 110.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material o-Fluorotoluene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-428-5
RTECSNumber XT2579000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.