Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzamidine Hydrochloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. F095925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F095925G 25 g
F09595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F095925G Supplier TCI America Supplier No. F095925G

Chemical Identifiers

CAS 456-14-4
Molecular Formula C7H8ClFN2
Molecular Weight (g/mol) 174.60
MDL Number MFCD04114421
InChI Key JQDATBKJKUWNGA-UHFFFAOYSA-N
Synonym 4-fluorobenzamidine hydrochloride, 4-fluorobenzimidamide hydrochloride, 4-fluorobenzamidine hcl, 4-fluoro-benzamidine hydrochloride, 4-fluorobenzenecarboximidamide hydrochloride, 4-fluorobenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-fluoro-, monohydrochloride, 4-fluorobenzimidamidehydrochloride, 4-fluorobenzamidine hydrogen chloride, 4-fluoranylbenzenecarboximidamide hydrochloride
PubChem CID 12456160
IUPAC Name hydrogen 4-fluorobenzene-1-carboximidamide chloride
SMILES [H+].[Cl-].NC(=N)C1=CC=C(F)C=C1

Specifications

Melting Point 211°C
Color White
Quantity 25 g
Formula Weight 174.60
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzamidine Hydrochloride Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.