Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzohydrazide 98.0+%, TCI America™
SDP

Catalog No. F054025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F054025G 25 g
F05405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F054025G Supplier TCI America Supplier No. F054025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 456-06-4
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.144
InChI Key UIVXXFYJRYVRKJ-UHFFFAOYSA-N
Synonym 4-fluorobenzhydrazide, 4-fluorobenzoic hydrazide, p-fluorobenzoic acid hydrazide, p-fluorobenzoylhydrazine, 4-fluorobenzoic acid hydrazide, hydrazine, p-fluorobenzoyl, 4-fluoro benzoyl hydrazine, benzoic acid, p-fluoro-, hydrazide, 4-fluorobenzenecarbohydrazide, 4-fluoro-benzoic acid hydrazide
PubChem CID 9972
IUPAC Name 4-fluorobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)F

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 25 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzohydrazide
RTECSNumber DH1310000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.