Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzoic hydrazide, 96%, Thermo Scientific™

Catalog No. p-4228664
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 456-06-4
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.14
InChI Key UIVXXFYJRYVRKJ-UHFFFAOYSA-N
Synonym 4-fluorobenzhydrazide, 4-fluorobenzoic hydrazide, p-fluorobenzoic acid hydrazide, p-fluorobenzoylhydrazine, 4-fluorobenzoic acid hydrazide, hydrazine, p-fluorobenzoyl, 4-fluoro benzoyl hydrazine, benzoic acid, p-fluoro-, hydrazide, 4-fluorobenzenecarbohydrazide, 4-fluoro-benzoic acid hydrazide
PubChem CID 9972
IUPAC Name 4-fluorobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)F

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 161°C to 164°C
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 154.14
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzoic hydrazide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.