Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzoylacetonitrile, 97%, Thermo Scientific™

Catalog No. AC429960050
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 4640-67-9
Molecular Formula C9H6FNO
Molecular Weight (g/mol) 163.15
MDL Number MFCD00662062
InChI Key LOJBBLDAJBJVBZ-UHFFFAOYSA-N
Synonym 4-fluorobenzoylacetonitrile, 3-4-fluorophenyl-3-oxopropanenitrile, p-fluorobenzoylacetonitrile, unii-k1ltb8k2dh, 3-4-fluorophenyl-3-oxopropionitrile, k1ltb8k2dh, 3-4-fluoro-phenyl-3-oxo-propionitrile, benzenepropanenitrile, 4-fluoro-beta-oxo, alpha-cyano-4-fluoroacetophenone, acetonitrile, p-fluorobenzoyl
PubChem CID 2783172
IUPAC Name 3-(4-fluorophenyl)-3-oxopropanenitrile
SMILES FC1=CC=C(C=C1)C(=O)CC#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 86.0°C to 87.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 163.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzoylacetonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.