Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzyl bromide, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
459-46-1
C7H6BrF
189.03
MFCD00000359
NVNPLEPBDPJYRZ-UHFFFAOYSA-N
4-fluorobenzyl bromide,1-bromomethyl-4-fluorobenzene,p-fluorobenzyl bromide,4-fluorobenzylbromide,benzene, 1-bromomethyl-4-fluoro,alpha-bromo-p-fluorotoluene,alpha-bromo-4-fluorotoluene,1-bromomethyl-4-fluoro-benzene,4-bromomethyl-1-fluorobenzene,para-fluorobenzyl bromide
68021
1-(bromomethyl)-4-fluorobenzene
C1=CC(=CC=C1CBr)F
459-46-1
C7H6BrF
189.03
MFCD00000359
NVNPLEPBDPJYRZ-UHFFFAOYSA-N
4-fluorobenzyl bromide,1-bromomethyl-4-fluorobenzene,p-fluorobenzyl bromide,4-fluorobenzylbromide,benzene, 1-bromomethyl-4-fluoro,alpha-bromo-p-fluorotoluene,alpha-bromo-4-fluorotoluene,1-bromomethyl-4-fluoro-benzene,4-bromomethyl-1-fluorobenzene,para-fluorobenzyl bromide
68021
1-(bromomethyl)-4-fluorobenzene
C1=CC(=CC=C1CBr)F
Specific Gravity 1.517
Refractive Index 1.5464 to 1.5484
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Density 1.5170g/mL
Flash Point >110°C
Quantity 5g
Boiling Point 85.0°C (15.0 mmHg)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula FC6H4CH2Br
Beilstein 05, II, 238
Formula Weight 189.03
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Fluorobenzyl bromide, 97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-291-5

TSCA : TSCA