Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[(4-Fluorophenoxy)methyl]oxirane, 97%, Thermo Scientific™

Catalog No. AC277130025
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18123-82-5
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00098471
InChI Key CGWDABYOHPEOAD-UHFFFAOYSA-N
Synonym 2-4-fluorophenoxy methyl oxirane, 2-4-fluorophenoxymethyl oxirane, 2-4-fluoro-phenoxymethyl-oxirane, oxirane,2-4-fluorophenoxy methyl, 4-fluorophenoxy methyl oxirane, 2-4-fluoranylphenoxy methyl oxirane, 4-fluoro-1-oxiran-2-ylmethoxy benzene, oxirane, 4-fluorophenoxy methyl, acmc-20m7dp
PubChem CID 2724792
IUPAC Name 2-[(4-fluorophenoxy)methyl]oxirane
SMILES C1C(O1)COC2=CC=C(C=C2)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 19.5°C
Color Colorless to Yellow
Boiling Point 79.0°C (1.0 Mbar)
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5060 to 1.5090
Quantity 2.5 g
Formula Weight 168.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material [(4-Fluorophenoxy)methyl]oxirane, 97%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.