Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenyl isocyanide, 95%

Catalog No. AC372620050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

CAS: 24075-34-1 | C7H4FN | 121.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24075-34-1
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD04117575
InChI Key FEUHGXVGNQTPGQ-UHFFFAOYSA-N
Synonym 4-fluorophenyl isocyanide, 4-fluorophenylisocyanide, 1-isocyano-4-fluorobenzene, benzene,1-fluoro-4-isocyano, benzene, 1-fluoro-4-isocyano-9ci
PubChem CID 2734801
IUPAC Name 1-fluoro-4-isocyanobenzene
SMILES FC1=CC=C(C=C1)[N+]#[C-]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown to Yellow
Density 1.1900g/mL
Boiling Point 62°C to 68°C (20.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4NC
Quantity 5 g
Specific Gravity 1.19
Solubility Information Solubility in water: insoluble
Formula Weight 121.11
Percent Purity 95%
Physical Form Liquid or Solid
Chemical Name or Material 4-Fluorophenyl isocyanide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.