Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. F064425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F064425G 25 g
F06445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F064425G Supplier TCI America Supplier No. F064425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1544-68-9
Molecular Formula C7H4FNS
Molecular Weight (g/mol) 153.174
MDL Number MFCD00004809
InChI Key NFIUJHJMCQQYDL-UHFFFAOYSA-N
Synonym 4-fluorophenyl isothiocyanate, p-fluorophenyl isothiocyanate, benzene, 1-fluoro-4-isothiocyanato, 4-fluorophenylisothiocyanate, 1-fluoro-4-isothiocyanato-benzene, isothiocyanic acid, p-fluorophenyl ester, isothiocyanic acid 4-fluorophenyl ester, 4-fluorobenzenisothiocyanate, 4-fluoroisothiocyanatobenzene, pubchem5008
PubChem CID 15241
IUPAC Name 1-fluoro-4-isothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)F

Specifications

Color Yellow
Boiling Point 228°C
Quantity 25 g
UN Number 2922
Formula Weight 153.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorophenyl Isothiocyanate
RTECSNumber NX8760000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.